Tauroursodeoxycholate dihydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 3α,7β-Dihydroxy-5β-cholan-24-oic acid N-(2-sulphoethyl)amide dihydrate, Tauroursodeoxycholic acid dihydrate, 2-((R)-4-((3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonic acid dihydrate

T1567-5G T1567-1G
TCIAT1567-5GEA 1320 DKK
TCIAT1567-5G TCIAT1567-1G
Tauroursodeoxycholate dihydrate ≥98.0% (by titrimetric analysis)
Tauroursodeoxycholate dihydrate
Formel: C₂₆H₄₅NNaO₆S·2H₂O
MDL Number: MFCD00069496
CAS-nummer: 14605-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=689d90dbc8a07d084073ce2e82daaa5d5048cbc5&vid=72504d987f04c2607ce0f4f7d90efc65b12837fb&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +42.0 to +48.0 deg (C=1, EtOH)
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR