1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Phenyl-1,2,3,6-tetrahydropyridine Hydrochloride, 4-Phenyl-1,2,3,6-tetrahydropyridine hydrochloride

T1356-25G T1356-5G
TCIAT1356-25GEA 1380 DKK
TCIAT1356-25G TCIAT1356-5G
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride ≥98.0% (by HPLC, titration analysis)
1,2,3,6-Tetrahydro-4-phenylpyridine hydrochloride
Formel: C₁₁H₁₃N
MDL Number: MFCD00012752
CAS-nummer: 43064-12-6
EINECS: 256-071-5
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 201.0 to 205.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR