Tetrabutylammonium thiocyanate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Tetrabutylammonium Rhodanide

T1278-25G T1278-250G
TCIAT1278-25GEA 583 DKK
TCIAT1278-25G TCIAT1278-250G
Tetrabutylammonium thiocyanate ≥97.0% (by titrimetric analysis)
Tetrabutylammonium thiocyanate
Formel: C₁₇H₃₆N₂S
MDL Number: MFCD00012140
CAS-nummer: 3674-54-2
EINECS: 222-943-9

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 119.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR