Tetrachloroterephthalonitrile ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

T1058-100MG T1058-25G T1058-5G
TCIAT1058-100MGEA 132 DKK
TCIAT1058-100MG TCIAT1058-25G TCIAT1058-5G
Tetrachloroterephthalonitrile ≥98.0% (by GC)
Tetrachloroterephthalonitrile
Formel: C₈Cl₄N₂
MDL Number: MFCD00059583
CAS-nummer: 1897-41-2
EINECS: 401-550-8
UN nummer: 3077
ADR: 9,III

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR