Dimethyl-3,3'-thiodipropionate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Dimethyl 3,3-Thiodipropionate, 3,3'-Thiodipropionic Acid Dimethyl Ester

T0901-25ML T0901-500ML
TCIAT0901-25MLEA 132 DKK
TCIAT0901-25ML TCIAT0901-500ML
Dimethyl-3,3'-thiodipropionate ≥98.0% (by GC)
Dimethyl-3,3'-thiodipropionate
Formel: C₈H₁₄O₄S
Molekylvægt: 206.26 g/mol
Kogepunkt: 164°C
MDL Number: MFCD00008465
CAS-nummer: 4131-74-2
EINECS: 223-948-9

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR