n-Octyl-p-toluenesulphonate ≥96.0% (by GC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Octyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid n-Octyl Ester
T0885-25G
T0885-500G
TCIAT0885-25GEA
624
DKK
TCIAT0885-25G
TCIAT0885-500G
n-Octyl-p-toluenesulphonate ≥96.0% (by GC, titration analysis)
n-Octyl-p-toluenesulphonate
Formel:
C₁₅H₂₄O₃S Molekylvægt: 284.41 g/mol |
MDL Number:
MFCD00059447 CAS-nummer: 3386-35-4 |
Specifikationer Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Ester Value) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...