Hexyl-p-toluenesulphonate ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
T0883-25G
T0883-500G
TCIAT0883-25GEA
386
DKK
TCIAT0883-25G
TCIAT0883-500G
Hexyl-p-toluenesulphonate ≥98.0% (by GC)
Hexyl-p-toluenesulphonate
Formel:
C₁₃H₂₀O₃S Kogepunkt: 197°C |
MDL Number:
MFCD00059439 CAS-nummer: 3839-35-8 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...