Tetrahydrofurfuryl butyrate 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Butyric Acid Tetrahydrofurfuryl Ester, 2-(Butyryloxymethyl)tetrahydrofuran

T0878-25ML T0878-500ML
TCIAT0878-25MLEA 427 DKK
TCIAT0878-25ML TCIAT0878-500ML
Tetrahydrofurfuryl butyrate 98,0 GC_ASSAY_METHOD
Tetrahydrofurfuryl butyrate
Formel: C₉H₁₆O₃
Molekylvægt: 172.22 g/mol
Kogepunkt: 227°C
Flammepunkt: 99°C
MDL Number: MFCD00059734
CAS-nummer: 2217-33-6
EINECS: 218-710-6

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR