TES (2-(Tris(hydroxymethyl)methylamino)ethan-1-sulfonsyre) ≥98.0% (by titrimetric analysis) good's buffer component for biological research

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

T0683-25G T0683-250G
TCIAT0683-25GEA 583 DKK
TCIAT0683-25G TCIAT0683-250G
TES (2-(Tris(hydroxymethyl)methylamino)ethan-1-sulfonsyre) ≥98.0% (by titrimetric analysis) good's buffer component for biological research
TES (2-(Tris(hydroxymethyl)methylamino)ethan-1-sulfonsyre)
Formel: C₆H₁₅NO₆S
Molekylvægt: 229.25 g/mol
MDL Number: MFCD00007532
CAS-nummer: 7365-44-8
EINECS: 230-906-3
Merck Index: 14,09176

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Absorbance max. 0.015 (0.1mol/L,H2O,260nm)
Solubility in Water almost transparency
Heavy metals (as Pb) max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR