Trimethylhexamethylenediamine (2,2,4- and 2,4,4- mixture) ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0600-25ML T0600-500ML
TCIAT0600-25MLEA 148 DKK
TCIAT0600-25ML TCIAT0600-500ML
Trimethylhexamethylenediamine (2,2,4- and 2,4,4- mixture) ≥99.0% (by GC, titration analysis)
Trimethylhexamethylenediamine (2,2,4- and 2,4,4- mixture)
Formel: C₉H₂₂N₂
Kogepunkt: 232°C
Flammepunkt: 127°C
MDL Number: MFCD00059849
CAS-nummer: 25513-64-8
EINECS: 247-063-2
UN nummer: 2327
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 % (total of isomers)
Purity (Nonaqueous Titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR