Trimethylene sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

T0474-25ML T0474-5ML
TCIAT0474-25MLEA 3220 DKK
TCIAT0474-25ML TCIAT0474-5ML
Trimethylene sulphide ≥98.0% (by GC)
Trimethylene sulphide
Formel: C₃H₆S
Kogepunkt: 94°C
Flammepunkt: –2°C
MDL Number: MFCD00042751
CAS-nummer: 287-27-4
EINECS: 206-015-0
UN nummer: 1993
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR