Thiamin hydrochlorid ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

T0181-25G T0181-500G T0181-100G
TCIAT0181-25GEA 197 DKK
TCIAT0181-25G TCIAT0181-500G TCIAT0181-100G
Thiamin hydrochlorid ≥98.0% (by HPLC, titration analysis)
Thiamin hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Molekylvægt: 337.27 g/mol
Smeltepunkt: 252
MDL Number: MFCD00012780
CAS-nummer: 67-03-8
EINECS: 200-641-8
Merck Index: 14,09295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee93060d98b444572fb217113adc013a1f2d0da6&vid=ee93060d98b444572fb217113adc013a1f2d0da6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR