Tetraphenylmethan ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T0169-1G T0169-5G
TCIAT0169-1GEA 476 DKK
TCIAT0169-1G TCIAT0169-5G
Tetraphenylmethan ≥96.0% (by GC)
Tetraphenylmethan
Formel: C₂₅H₂₀
Molekylvægt: 320.43 g/mol
Kogepunkt: 431°C
Smeltepunkt: 283°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00014428
CAS-nummer: 630-76-2
EINECS: 211-144-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6695c9b0be083749909a4cb14b3a204e85b81201&vid=6695c9b0be083749909a4cb14b3a204e85b81201&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR