Leucoquinizarin ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Anthracene-1,4,9,10-tetraol, leuco-Quinizarin, 1,4-Dihydroxyanthrahydroquinone, 1,4,9,10-Tetrahydroxyanthracene, 2,3-Dihydro-9,10-dihydroxy-1,4-anthraquinone

T0116-25G T0116-500G
TCIAT0116-25GEA 444 DKK
TCIAT0116-25G TCIAT0116-500G
Leucoquinizarin ≥98.0% (by HPLC)
Leucoquinizarin
Formel: C₁₄H₁₀O₄
Molekylvægt: 242.23 g/mol
MDL Number: MFCD00058948
CAS-nummer: 476-60-8
EINECS: 207-507-8

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR