Dibutyl-L-(+)-tartrate 98,0 GC_ASSAY_METHOD

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (2R,3R)-Dibutyl 2,3-dihydroxysuccinate, L-(+)-Tartaric acid dibutyl ester

T0005-25G T0005-500G
TCIAT0005-25GEA 238 DKK
TCIAT0005-25G TCIAT0005-500G
Dibutyl-L-(+)-tartrate 98,0 GC_ASSAY_METHOD
Dibutyl-L-(+)-tartrate
Formel: C₁₂H₂₂O₆
Molekylvægt: 262.30 g/mol
Flammepunkt: 160°C
MDL Number: MFCD00009443
CAS-nummer: 87-92-3
EINECS: 201-784-9

Bestil nu

Specifikationer Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +11.0 to +14.0 deg (C=5, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR