Sodium methacrylate ≥98.0% (by titrimetric analysis) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
S0987-25G
S0987-100G
TCIAS0987-25GEA
255
DKK
TCIAS0987-25G
TCIAS0987-100G
Sodium methacrylate ≥98.0% (by titrimetric analysis) stabilised
Sodium methacrylate
Formel:
C₄H₅NaO₂ |
MDL Number:
MFCD00045886 CAS-nummer: 5536-61-8 EINECS: 226-896-5 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Bromination Method) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...