9,9'-Spirobi[9H-9-silafluorene] ≥96.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

S0983-200MG S0983-1G
TCIAS0983-200MGEA 337 DKK
TCIAS0983-200MG TCIAS0983-1G
9,9'-Spirobi[9H-9-silafluorene] ≥96.0% (by GC)
9,9'-Spirobi[9H-9-silafluorene]
Formel: C₂₄H₁₆Si
Kogepunkt: 460°C
MDL Number: MFCD00093804
CAS-nummer: 159-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f8e62aafc55568821c55c83cb9685b13ebbf76b&vid=0f8e62aafc55568821c55c83cb9685b13ebbf76b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 230.0 to 234.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR