Sodium-2-propylvalerate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

S0894-25G S0894-100G
TCIAS0894-25GEA 517 DKK
TCIAS0894-25G TCIAS0894-100G
Sodium-2-propylvalerate ≥98.0% (by titrimetric analysis)
Sodium-2-propylvalerate
Formel: C₈H₁₅O₂Na
MDL Number: MFCD00078604
CAS-nummer: 1069-66-5
EINECS: 213-961-8
Merck Index: 14,09913
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c39a6e2138be203c76120d476025a686de2ccba&vid=0c39a6e2138be203c76120d476025a686de2ccba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR