N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
7-Methoxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, 2,5-Dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate
S0867-1G
S0867-100MG
TCIAS0867-1GEA
2320
DKK
TCIAS0867-1G
TCIAS0867-100MG
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥98.0% (by HPLC, total nitrogen)
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate
Formel:
C₁₅H₁₁NO₇ Molekylvægt: 317.25 g/mol Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00467593 CAS-nummer: 150321-92-9 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...