Sulphamethazine sodium salt ≥97.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide sodium salt, Sodium ((4-aminophenyl)sulphonyl)(4,6-dimethylpyrimidin-2-yl)amide, Sulphadimidine sodium salt, Sodium sulphamethazine
S0583-25G
S0583-500G
TCIAS0583-25GEA
132
DKK
TCIAS0583-25G
TCIAS0583-500G
Sulphamethazine sodium salt ≥97.0% (by HPLC, total nitrogen)
Sulphamethazine sodium salt
Formel:
C₁₂H₁₃N₄NaO₂S Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00068333 CAS-nummer: 1981-58-4 EINECS: 217-840-0 Merck Index: 14,08915 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...