Stanozolol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

S0530-1G S0530-5G
TCIAS0530-1GEA 476 DKK
TCIAS0530-1G TCIAS0530-5G
Stanozolol ≥98.0% (by HPLC, titration analysis)
Stanozolol
Formel: C₂₁H₃₂N₂O
Smeltepunkt: 242°C
MDL Number: MFCD00133084
CAS-nummer: 10418-03-8
EINECS: 233-894-8
Merck Index: 14,08794

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on dried substance)
Specific rotation [a]20/D +31.0 to +37.0 deg (C=0.4, CHCl3)
Drying loss max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR