2,3-Dimercapto-1-propanesulphonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: DL-2,3-Dimercapto-1-propanesulphonic acid, sodium salt monohydrate, DMPS, Sodium 2,3-dimercaptopropane-1-sulphonate hydrate, 2,3-Dimercaptopropanesulphonic acid sodium salt, Sodium 2,3-Dimercaptopropanesulphonate monohydrate

S0524-25G S0524-5G
TCIAS0524-25GEA 2210 DKK
TCIAS0524-25G TCIAS0524-5G
2,3-Dimercapto-1-propanesulphonic acid sodium salt monohydrate ≥97.0% (by titrimetric analysis)
2,3-Dimercapto-1-propanesulphonic acids odium salt monohydrate
Formel: C₃H₉NaO₄S₃·H₂O
Molekylvægt: 228.30 g/mol (210.28 anhy)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00149543
CAS-nummer: 207233-91-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 97.0 % (calcd.on anh.substance)

Specifikationer

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR