4-Sulphocalix[6]arene hydrate ≥95.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Hexahydroxycalix[6]arenehexasulphonic acid, Hexasulpho(hexahydroxy)calix[6]arene
S0470-1G
S0470-5G
TCIAS0470-1GEA
1470
DKK
TCIAS0470-1G
TCIAS0470-5G
4-Sulphocalix[6]arene hydrate ≥95.0% (by HPLC)
4-Sulphocalix[6]arene hydrate
Formel:
C₄₂H₃₆O₂₄S₆ |
MDL Number:
MFCD00143376 CAS-nummer: 102088-39-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Water | 3.0 to 15.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...