Vinyl stearate ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Stearic Acid Vinyl Ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
S0241-25G
S0241-100G
TCIAS0241-25GEA
181
DKK
TCIAS0241-25G
TCIAS0241-100G
Vinyl stearate ≥95.0% (by GC) stabilised
Vinyl stearate
Formel:
C₂₀H₃₈O₂ Molekylvægt: 310.52 g/mol |
MDL Number:
MFCD00026667 CAS-nummer: 111-63-7 EINECS: 203-890-0 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...