Succinaldehyde disodium bisulphite ≥92.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

S0216-25G S0216-500G
TCIAS0216-25GEA 255 DKK
TCIAS0216-25G TCIAS0216-500G
Succinaldehyde disodium bisulphite ≥92.0% (by titrimetric analysis)
Succinaldehyde disodium bisulphite
Formel: C₄H₁₀O₈S₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00024982
CAS-nummer: 5450-96-4
EINECS: 226-676-9

Bestil nu

Specifikationer Testresultater

Appearance White powder to crystal
Purity (Iodometric Titration) min. 92.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 12.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR