Stearonitrile ≥92.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Octadecanenitrile, Heptadecyl Cyanide

S0085-25G S0085-5G
TCIAS0085-25GEA 1290 DKK
TCIAS0085-25G TCIAS0085-5G
Stearonitrile ≥92.0% (by GC)
Stearonitrile
Formel: C₁₈H₃₅N
MDL Number: MFCD00001947
CAS-nummer: 638-65-3
EINECS: 211-345-3

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 92.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR