Roflumilast N-oxide ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3,5-Dichloro-4-[3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzamido]pyridine 1-Oxide, 3,5-Dichloro-4-(3-(cyclopropylmethoxy)-4-(difluoromethoxy)benzamido)pyridine 1-oxide
R0217-25MG
R0217-5MG
TCIAR0217-25MGEA
1550
DKK
TCIAR0217-25MG
TCIAR0217-5MG
Roflumilast N-oxide ≥98.0% (by HPLC)
Roflumilast N-oxide
Formel:
C₁₇H₁₄Cl₂F₂N₂O₄ Opbevaringstemperatur: Fryser |
MDL Number:
MFCD04112984 CAS-nummer: 292135-78-5 |
Specifikationer Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 6.00 to 7.00 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...