Ruthenium(III) chloride oxide

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Ruthenium oxychloride, Tetraamminechlorohydroxoruthenium(III) chloride, Ruthenium Red

R0204-1G R0204-5G
TCIAR0204-1GEA 1870 DKK
TCIAR0204-1G TCIAR0204-5G
Ruthenium(III) chloride oxide
Ruthenium(III) chloride oxide
Formel: H₄₂Cl₆N₁₄O₂Ru₃
Molekylvægt: 786.354 g/mol
MDL Number: MFCD00011479
CAS-nummer: 11103-72-3

Bestil nu

Specifikationer Testresultater

Appearance Gray to Dark purple to Black powder to crystal
Lambda max. 532.0 to 536.0 nm (0.1mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 250 (0.1mol/L AcONH4 sol., 532.0 to 536.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR