Rifamycin S ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
R0200-5G
R0200-1G
TCIAR0200-5GEA
3370
DKK
TCIAR0200-5G
TCIAR0200-1G
Rifamycin S ≥98.0% (by HPLC)
Rifamycin S
Formel:
C₃₇H₄₅NO₁₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD06198807 CAS-nummer: 13553-79-2 EINECS: 236-938-4 Merck Index: 14,08217 |
Specifikationer Testresultater
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Specific rotation [a]20/D | +451 to +500 deg (C=0.1, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...