R-(-)-Rolipram ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (R)-(-)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]pyrrolidin-2-one, R-(-)-4-[3-(Cyclopentyloxy)-4-methoxyphenyl]-2-pyrrolidinone

R0182-10MG R0182-50MG
TCIAR0182-10MGEA 591 DKK
TCIAR0182-10MG TCIAR0182-50MG
R-(-)-Rolipram ≥98.0% (by HPLC)
R-(-)-Rolipram
Formel: C₁₆H₂₁NO₃
Molekylvægt: 275.34 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD03093861
CAS-nummer: 85416-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b471b5ab3ad67b5bfe3bc4ed0dc7f46e894478c3&vid=b471b5ab3ad67b5bfe3bc4ed0dc7f46e894478c3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Elemental analysis (Nitrogen) 4.50 to 5.50 %
Melting point 132.0 to 136.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR