Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Ru(OAc)2[(S)-binap]
R0167-200MG
R0167-1G
TCIAR0167-200MGEA
304
DKK
TCIAR0167-200MG
TCIAR0167-1G
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Formel:
C₄₈H₃₈O₄P₂Ru |
MDL Number:
MFCD09753020 CAS-nummer: 261948-85-0 |
|
Specifikationer Testresultater
Appearance | Yellow to Amber to Dark purple powder to crystal |
IR | to pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...