(R)-RUCY®-XylBINAP

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: RuCl[(R)-daipena][(R)-xylbinap]

R0139-200MG R0139-1G
TCIAR0139-200MGEA 353 DKK
TCIAR0139-200MG TCIAR0139-1G
(R)-RUCY®-XylBINAP
(R)-RUCY®-XylBINAP
Formel: C₇₁H₇₃ClN₂O₂P₂Ru
MDL Number: MFCD20261161
CAS-nummer: 1384974-38-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c08105a4cfe7b50a6ecb755ce8a0d56f3c04341f&vid=c08105a4cfe7b50a6ecb755ce8a0d56f3c04341f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Yellow to Brown to Dark green powder to crystal
Content (Ruthenium) 8 to 9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR