Ru-MACHO® (contains 5% Toluene at maximum)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

R0136-200MG R0136-1G
TCIAR0136-200MGEA 296 DKK
TCIAR0136-200MG TCIAR0136-1G
Ru-MACHO® (contains 5% Toluene at maximum)
Ru-MACHO®
Formel: C₂₉H₃₀ClNOP₂Ru
MDL Number: MFCD22377801
CAS-nummer: 1295649-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9d7bfa2003ed67acc23c38f678a09fb54e97da5&vid=a9d7bfa2003ed67acc23c38f678a09fb54e97da5&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Content (Ruthenium) 15 to 17 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR