Raclopride ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

R0094-200MG R0094-1G
TCIAR0094-200MGEA 862 DKK
TCIAR0094-200MG TCIAR0094-1G
Raclopride ≥98.0% (by HPLC, total nitrogen)
Raclopride
Formel: C₁₅H₂₀Cl₂N₂O₃
MDL Number: MFCD02179386
CAS-nummer: 84225-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=278fabcc4b674b8ca2bd01a81cd9a9563c3272dc&vid=278fabcc4b674b8ca2bd01a81cd9a9563c3272dc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light red to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Specific rotation [a]20/D -67.0 to -71.0 deg (C=2,Acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR