Quinoline-N-oxide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Quinoline 1-oxide, Quinoline-1-oxide

Q0104-25G Q0104-5G
TCIAQ0104-25GEA 1240 DKK
TCIAQ0104-25G TCIAQ0104-5G
Quinoline-N-oxide ≥98.0% (by GC)
Quinoline-N-oxide
Formel: C₉H₇NO
Molekylvægt: 145.161 g/mol
Kogepunkt: 172°C
MDL Number: MFCD00006737
CAS-nummer: 1613-37-2
EINECS: 216-560-6

Bestil nu

Specifikationer Testresultater

Appearance White to Gray to Brown powder to crystaline
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR