(8-Quinolinolato)lithium ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Lithium quinolin-8-olate, Liq

Q0100-1G Q0100-5G
TCIAQ0100-1GEA 255 DKK
TCIAQ0100-1G TCIAQ0100-5G
(8-Quinolinolato)lithium ≥98.0% (by HPLC, titration analysis)
(8-Quinolinolato)lithium
Formel: C₉H₆LiNO
Molekylvægt: 151.09 g/mol
MDL Number: MFCD00152778
CAS-nummer: 25387-93-3
EINECS: 246-932-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5f24c442546b77c641d093e7d279169c6c688d7&vid=b5f24c442546b77c641d093e7d279169c6c688d7&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR