2-(Propylamino)ethanol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: N-Propylethanolamine

P2743-25G P2743-5G
TCIAP2743-25GEA 2250 DKK
TCIAP2743-25G TCIAP2743-5G
2-(Propylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(Propylamino)ethanol
Formel: C₅H₁₃NO
Molekylvægt: 103.16 g/mol
Kogepunkt: 182°C
Massefylde: 0.901
Flammepunkt: 78°C
MDL Number: MFCD00002844
CAS-nummer: 16369-21-4
EINECS: 240-426-6
UN nummer: 2735
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR