Polyethylene glycol methyl ether acrylate n=∼9 stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
mPEG-acrylate, Polyethylene glycol monomethyl ether acrylate
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
P2698-25G
P2698-500G
TCIAP2698-25GEA
279
DKK
TCIAP2698-25G
TCIAP2698-500G
Polyethylene glycol methyl ether acrylate n=∼9 stabilised
Polyethylene glycol methyl ether acrylate
Formel:
CH₃(OCH₂CH₂)nOCOC=CH₂ |
MDL Number:
MFCD00803685 CAS-nummer: 32171-39-4 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
The average value of n | 8.5 to 9.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...