trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-ol, trans-4-(trans-4-Amylcyclohexyl)cyclohexanol, trans,trans-4'-Amyl-[1,1'-bi(cyclohexane)]-4-ol, trans,trans-4'-Pentyl-[1,1'-bi(cyclohexane)]-4-ol

P2690-25G P2690-5G
TCIAP2690-25GEA 1800 DKK
TCIAP2690-25G TCIAP2690-5G
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol ≥98.0% (by GC)
trans-4-(trans-4-Pentylcyclohexyl)cyclohexanol
Formel: C₁₇H₃₂O
Molekylvægt: 252.44 g/mol
MDL Number: MFCD09752070
CAS-nummer: 82575-70-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 124.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR