Potassium trifluoro(2-fluoropyridin-3-yl)borate ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Potassium 2-Fluoropyridine-3-trifluoroborate

P2675-1G P2675-5G
TCIAP2675-1GEA 632 DKK
TCIAP2675-1G TCIAP2675-5G
Potassium trifluoro(2-fluoropyridin-3-yl)borate ≥98.0% (by HPLC, titration analysis)
Potassium trifluoro(2-fluoropyridin-3-yl)borate
Formel: C₅H₃BF₄NK
MDL Number: MFCD09993002
CAS-nummer: 1111732-91-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR