4-P-PDOT ≥95.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2587-25MG P2587-5MG
TCIAP2587-25MGEA 2060 DKK
TCIAP2587-25MG TCIAP2587-5MG
4-P-PDOT ≥95.0% (by GC)
4-P-PDOT
Formel: C₁₉H₂₁NO
MDL Number: MFCD00901545
CAS-nummer: 134865-74-0

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 164.0 to 168.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR