2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (9,9'-Spirobi[fluoren]-3-yl)boronic acid Pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluorene]

P2562-200MG P2562-1G
TCIAP2562-200MGEA 682 DKK
TCIAP2562-200MG TCIAP2562-1G
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₃₁H₂₇BO₂
MDL Number: MFCD30187274
CAS-nummer: 1346007-05-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d471ff8d028064809899766b91b893a11654c9b&vid=4d471ff8d028064809899766b91b893a11654c9b&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 214.0 to 218.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR