1-Pyrrolidinamine ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
1-Aminopyrrolidine
P2560-250MG
P2560-1G
TCIAP2560-250MGEA
1460
DKK
TCIAP2560-250MG
TCIAP2560-1G
1-Pyrrolidinamine ≥98.0% (by titrimetric analysis)
1-Pyrrolidinamine
Formel:
C₄H₁₀N₂ Kogepunkt: 30°C |
MDL Number:
MFCD07368381 CAS-nummer: 16596-41-1 UN nummer: 2734 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...