10-Phenyl-9(10H)-acridone ≥98.0% (by HPLC)
Leverandør: TCI
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: 10-Phenylacridin-9(10H)-one
P2537-1G P2537-5G
TCIAP2537-1GEA 641 DKK
TCIAP2537-1G TCIAP2537-5G
10-Phenyl-9(10H)-acridone ≥98.0% (by HPLC)
10-Phenyl-9(10H)-acridone
Formel: C₁₉H₁₃NO
Molekylvægt: 271.32 g/mol
MDL Number: MFCD01463673
CAS-nummer: 5472-23-1
Bestil nu
Specifikationer Testresultater
Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 96.0 %