1-(2-Pyrimidinyl)-1H-indole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2505-1G P2505-5G
TCIAP2505-1GEA 1200 DKK
TCIAP2505-1G TCIAP2505-5G
1-(2-Pyrimidinyl)-1H-indole ≥98.0% (by GC)
1-(2-Pyrimidinyl)-1H-indole
Formel: C₁₂H₉N₃
MDL Number: MFCD28556906
CAS-nummer: 221044-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0536e9bbb1d17328e8b8fbe78c59b33262ed0493&vid=0536e9bbb1d17328e8b8fbe78c59b33262ed0493&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR