Kaliumdisulfit ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite

P2480-25G P2480-500G
TCIAP2480-25GEA 132 DKK
TCIAP2480-25G TCIAP2480-500G
Kaliumdisulfit ≥97.0% (by titrimetric analysis)
Potassium disulphite
Formel: K₂O₅S₂
Molekylvægt: 222.33 g/mol
Smeltepunkt: 190°C
MDL Number: MFCD00167605
CAS-nummer: 16731-55-8
EINECS: 240-795-3
Merck Index: 14,07645
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebb1876a2b8a3d9acca71314972f9dda08cafbcf&vid=ebb1876a2b8a3d9acca71314972f9dda08cafbcf&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR