PD-0173074 ≥95.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: PD 173074, 1-(tert-Butyl)-3-[7-[[4-(diethylamino)butyl]amino]-3-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-2-yl]urea, PD173074, 1-(tert-Butyl)-3-(2-((4-(diethylamino)butyl)amino)-6-(3,5-dimethoxyphenyl)pyrido[2,3-d]pyrimidin-7-yl)urea

P2474-10MG P2474-50MG
TCIAP2474-10MGEA 411 DKK
TCIAP2474-10MG TCIAP2474-50MG
PD-0173074 ≥95.0% (by HPLC)
PD-0173074
Formel: C₂₈H₄₁N₇O₃
Smeltepunkt: 86°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08705327
CAS-nummer: 219580-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3972f3de789629f579b32b13bc4282a8be07f7f&vid=d3972f3de789629f579b32b13bc4282a8be07f7f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 18.00 to 19.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR