trans,trans-4'-Pentyl-4-bicyclohexylmethanol ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
((trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-yl)methanol, trans,trans-4'-Amyl-4-bicyclohexylmethanol
P2433-1G
P2433-5G
TCIAP2433-1GEA
230
DKK
TCIAP2433-1G
TCIAP2433-5G
trans,trans-4'-Pentyl-4-bicyclohexylmethanol ≥98.0% (by GC)
trans,trans-4'-Pentyl-4-bicyclohexylmethanol
Formel:
C₁₈H₃₄O Molekylvægt: 266.47 g/mol |
MDL Number:
MFCD16036281 CAS-nummer: 82598-08-1 |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...