Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥93.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Phenyl(2,4,6-trimethoxyphenyl)iodonium Tosylate

P2412-200MG P2412-1G
TCIAP2412-200MGEA 534 DKK
TCIAP2412-200MG TCIAP2412-1G
Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥93.0% (by HPLC)
Phenyl(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate
Formel: C₂₂H₂₃IO₆S
Molekylvægt: 542.38 g/mol
Opbevaringstemperatur: Køleskab
CAS-nummer: 936326-60-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a10c2f77005e1b99aea8f97f72712eb089c4d131&vid=a10c2f77005e1b99aea8f97f72712eb089c4d131&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 93.0 % (total of Ion-Pair)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR