3-Phenyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Phenylisoxazole-5-carboxylic acid
P2390-1G
P2390-5G
TCIAP2390-1GEA
386
DKK
TCIAP2390-1G
TCIAP2390-5G
3-Phenyl-5-isoxazolecarboxylic acid ≥98.0% (by GC)
3-Phenyl-5-isoxazolecarboxylic acid
Formel:
C₁₀H₇NO₃ |
MDL Number:
MFCD01936010 CAS-nummer: 14442-12-7 |
|
Specifikationer Testresultater
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...